ENA375228077 Enamine

1-(bromomethyl)-1-(methanesulfonylmethyl)cyclopropane

  • MDL number MFCD20886228
  • Cas Number : 1423032-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50...52
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116017

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116017
Availability details for
Quantity
  • Availability
  • Add to Cart