ENA375228078 Enamine

1-bromo-3-(bromomethyl)-2-(difluoromethoxy)benzene

  • MDL number MFCD14649650
  • Cas Number : 1248106-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116018
Availability details for
Quantity
  • Availability
  • Add to Cart