ENA375228080 Enamine

1-(3,5-dimethylcyclohexyl)ethan-1-one

  • MDL number MFCD20404925
  • Cas Number : 1341316-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116023

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116023
Availability details for
Quantity
  • Availability
  • Add to Cart