ENA375228081 Enamine

6-bromo-2-methylpyridine-3-carboxylic acid

  • MDL number MFCD13188658
  • Cas Number : 1060805-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116024

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116024
Availability details for
Quantity
  • Availability
  • Add to Cart