ENA375228084 Enamine

tert-butyl N-(3-cyano-2-hydroxypropyl)carbamate

  • MDL number MFCD22565853
  • Cas Number : 120245-01-4
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-cyano-2-hydroxypropylcarbamate
Inchi Code 1S/C9H16N2O3/c1-9(2,3)14-8(13)11-6-7(12)4-5-10/h7,12H,4,6H2,1-3H3,(H,11,13)
Inchi Key IYEDXPKCFGTWDN-UHFFFAOYSA-N

Description

Purity 95
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-109363

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart