ENA375228090 Enamine

4-(4-methylpiperidin-1-yl)cyclohexan-1-amine

  • MDL number MFCD17220720
  • Cas Number : 1340279-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116070
Availability details for
Quantity
  • Availability
  • Add to Cart