ENA375228094 Enamine

N-[3-(methylamino)propyl]methanesulfonamide

  • MDL number MFCD12085389
  • Cas Number : 1154566-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116075
Availability details for
Quantity
  • Availability
  • Add to Cart