ENA375228098 Enamine

1-(4-bromobenzoyl)-4-(2-methoxyethyl)-1,4-diazepane

  • MDL number MFCD22566048
  • Cas Number : 1184489-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116080
Availability details for
Quantity
  • Availability
  • Add to Cart