ENA375228118 Enamine

1-methyl-1H-indazole-6-carbonitrile

  • MDL number MFCD12028576
  • Cas Number : 267413-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114547
Availability details for
Quantity
  • Availability
  • Add to Cart