ENA375228126 Enamine

2-(4-chlorophenyl)-2H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD11169395
  • Cas Number : 89522-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116130
Availability details for
Quantity
  • Availability
  • Add to Cart