ENA375228127 Enamine

4-(3-fluoro-4-methoxyphenyl)-1,2-dihydrophthalazin-1-one

  • MDL number MFCD13707711
  • Cas Number : 137382-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114548
Availability details for
Quantity
  • Availability
  • Add to Cart