ENA375228128 Enamine

2-cyclopropylethane-1-sulfonyl chloride

  • MDL number MFCD13190121
  • Cas Number : 1196155-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116132
Availability details for
Quantity
  • Availability
  • Add to Cart