ENA375228132 Enamine

2-(1,3-dimethyl-1H-pyrazol-4-yl)ethan-1-amine

  • MDL number MFCD08449792
  • Cas Number : 1007461-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114551
Availability details for
Quantity
  • Availability
  • Add to Cart