ENA375228137 Enamine

5-chloro-2-methyl-1H-indole-3-carboxylic acid

  • MDL number MFCD07776610
  • Cas Number : 889942-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193...195
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116166

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-116166
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart