ENA375228138 Enamine

N2,N2-dimethylpyrimidine-2,5-diamine

  • MDL number MFCD11656641
  • Cas Number : 56621-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-116167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116167
Availability details for
Quantity
  • Availability
  • Add to Cart