ENA375228140 Enamine

3-methylimidazo[2,1-b][1,3]thiazole-2-carboxylic acid

  • MDL number MFCD11889936
  • Cas Number : 1017273-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-116169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116169
Availability details for
Quantity
  • Availability
  • Add to Cart