ENA375228154 Enamine

2-(oxolan-3-yl)-1-phenylethan-1-amine

  • MDL number MFCD16858294
  • Cas Number : 1339699-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116187
Availability details for
Quantity
  • Availability
  • Add to Cart