ENA375228157 Enamine

ethyl 2-(4-bromo-3-fluorophenyl)acetate

  • MDL number MFCD19105270
  • Cas Number : 1296223-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116190

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116190
Availability details for
Quantity
  • Availability
  • Add to Cart