ENA375228164 Enamine

2-chloro-4-(morpholin-4-yl)-6-(2,2,2-trifluoroethoxy)-1,3,5-triazine

  • MDL number MFCD18032344
  • Cas Number : 1283693-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116205

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116205
Availability details for
Quantity
  • Availability
  • Add to Cart