ENA375228165 Enamine

2-(2-aminoethoxy)-1,1-dimethoxyethane

  • MDL number MFCD19215935
  • Cas Number : 1228258-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116206
Availability details for
Quantity
  • Availability
  • Add to Cart