ENA375228167 Enamine

1-(6-methylpyridine-2-carbonyl)piperidine-2-carboxylic acid

  • MDL number MFCD09740196
  • Cas Number : 1101831-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116216

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116216
Availability details for
Quantity
  • Availability
  • Add to Cart