ENA375228169 Enamine

4-bromo-3-hydroxybenzoic acid

  • MDL number MFCD08056371
  • Cas Number : 14348-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109810

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-109810
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart