ENA375228180 Enamine

1-(1,3-thiazol-2-yl)butane-1,3-dione

  • MDL number MFCD21120568
  • Cas Number : 1423028-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116256

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116256
Availability details for
Quantity
  • Availability
  • Add to Cart