ENA375228192 Enamine

3-(3,4-dichlorophenyl)-1,2-thiazole-5-carboxylic acid

  • MDL number MFCD21304978
  • Cas Number : 1423033-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208...210
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116279

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116279
Availability details for
Quantity
  • Availability
  • Add to Cart