ENA375228195 Enamine

N1,N1,3,5-tetramethylbenzene-1,4-diamine

  • MDL number MFCD09035943
  • Cas Number : 27746-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116284
Availability details for
Quantity
  • Availability
  • Add to Cart