ENA375228199 Enamine

6-chloro-[1,2,4]triazolo[1,5-a]pyridin-2-amine

  • MDL number MFCD11846610
  • Cas Number : 1239647-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183...185
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116301

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116301
Availability details for
Quantity
  • Availability
  • Add to Cart