ENA375228206 Enamine

2-bromo-1-(pyrrolidin-1-yl)ethan-1-one

  • MDL number MFCD06800371
  • Cas Number : 90892-09-4
  • Molecular Weight : 192.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 30-33
Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-116311

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P285,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P304+P341,P305+P351+P338,P310,P312,P330,P342+P311,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-116311
pictograms GHS05,GHS07,GHS08
hstatements H302,H314,H334,H335
Availability details for
Quantity
  • Availability
  • Add to Cart