ENA375228207 Enamine

1-bromo-4-(2-methylbutan-2-yl)benzene

  • MDL number MFCD00028024
  • Cas Number : 57263-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116312
Availability details for
Quantity
  • Availability
  • Add to Cart