ENA375228211 Enamine

1-[(3,4-dimethylphenyl)sulfanyl]butan-2-amine

  • MDL number MFCD20374158
  • Cas Number : 1339381-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116317
Availability details for
Quantity
  • Availability
  • Add to Cart