ENA375228222 Enamine

6-tert-butyl-1H,2H,3H-pyrazolo[1,5-a]imidazol-2-one

  • MDL number MFCD13193596
  • Cas Number : 663616-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-116372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116372
Availability details for
Quantity
  • Availability
  • Add to Cart