ENA375228225 Enamine

4-(morpholin-4-yl)-N-[(pyridin-2-yl)methyl]aniline

  • MDL number MFCD11058403
  • Cas Number : 940366-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116379
Availability details for
Quantity
  • Availability
  • Add to Cart