ENA375228226 Enamine

3-(2-chlorophenoxy)piperidine hydrochloride

  • MDL number MFCD12026501
  • Cas Number : 1185297-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116381

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116381
Availability details for
Quantity
  • Availability
  • Add to Cart