ENA375228233 Enamine

(1,2,3,4-tetrahydroquinolin-5-yl)urea

  • MDL number MFCD09811500
  • Cas Number : 1016712-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116390
Availability details for
Quantity
  • Availability
  • Add to Cart