ENA375228235 Enamine

2-[methyl(thiolan-3-yl)amino]acetic acid

  • MDL number MFCD11634233
  • Cas Number : 1095626-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116394

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116394
Availability details for
Quantity
  • Availability
  • Add to Cart