ENA375228238 Enamine

5-(chloromethyl)-3-cyclopentyl-1,2,4-oxadiazole

  • MDL number MFCD12807701
  • Cas Number : 1179864-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116397

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116397
Availability details for
Quantity
  • Availability
  • Add to Cart