ENA375228239 Enamine

1,2-difluoro-3-isothiocyanatobenzene

  • MDL number MFCD03093816
  • Cas Number : 363179-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116398

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-116398
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart