ENA375228245 Enamine

ethyl 2-[(1E)-cyclopropylidene]propanoate

  • MDL number MFCD19439678
  • Cas Number : 55281-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116413

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116413
Availability details for
Quantity
  • Availability
  • Add to Cart