ENA375228256 Enamine

1-{[cyclohexyl(methyl)carbamoyl]methyl}piperidine-2-carboxylic acid

  • MDL number MFCD13696089
  • Cas Number : 1218172-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114579

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114579
Availability details for
Quantity
  • Availability
  • Add to Cart