ENA375228258 Enamine

4-phenylpentanoic acid

  • MDL number MFCD00086322
  • Cas Number : 16433-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110188

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110188
Availability details for
Quantity
  • Availability
  • Add to Cart