ENA375228263 Enamine

1-(1-methyl-1H-pyrazol-4-yl)ethane-1,2-diamine

  • MDL number MFCD11503966
  • Cas Number : 1154875-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116505
Availability details for
Quantity
  • Availability
  • Add to Cart