ENA375228266 Enamine

4-ethyl-3-fluoroaniline

  • MDL number MFCD19205834
  • Cas Number : 1034497-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116509
Availability details for
Quantity
  • Availability
  • Add to Cart