ENA375228272 Enamine

5-[(4-bromo-1H-pyrazol-1-yl)methyl]-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD08700531
  • Cas Number : 1007514-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116518
Availability details for
Quantity
  • Availability
  • Add to Cart