ENA375228275 Enamine

(5-bromo-2,3-dihydro-1-benzofuran-2-yl)methanol

  • MDL number MFCD16037374
  • Cas Number : 197577-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63...65
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116523

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116523
Availability details for
Quantity
  • Availability
  • Add to Cart