ENA375228276 Enamine

N-{[2-(propan-2-yloxy)pyridin-4-yl]methyl}cyclopropanamine

  • MDL number MFCD16704416
  • Cas Number : 1250524-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116524
Availability details for
Quantity
  • Availability
  • Add to Cart