ENA375228279 Enamine

7-methoxy-2,3,4,5-tetrahydro-1H-2-benzazepin-1-one

  • MDL number MFCD00667644
  • Cas Number : 3648-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-158
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116527
Availability details for
Quantity
  • Availability
  • Add to Cart