ENA375228280 Enamine

N-cyclohexyl-2H-1,3-benzodioxol-5-amine

  • MDL number MFCD00981084
  • Cas Number : 176905-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116528

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116528
Availability details for
Quantity
  • Availability
  • Add to Cart