ENA375228282 Enamine

ethyl 2-(3,5-dichlorophenyl)-2-(ethylamino)acetate

  • MDL number MFCD12517952
  • Cas Number : 1218548-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116530
Availability details for
Quantity
  • Availability
  • Add to Cart