ENA375228288 Enamine

4-(bromomethyl)-4-(methanesulfonylmethyl)oxane

  • MDL number MFCD21181555
  • Cas Number : 1423034-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116538
Availability details for
Quantity
  • Availability
  • Add to Cart