ENA375228293 Enamine

3-(methylsulfanyl)pentanoic acid

  • MDL number MFCD17288057
  • Cas Number : 1310210-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116543

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116543
Availability details for
Quantity
  • Availability
  • Add to Cart