ENA375228298 Enamine

4-[(4-chlorophenyl)amino]benzonitrile

  • MDL number MFCD11125897
  • Cas Number : 1040022-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114621

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114621
Availability details for
Quantity
  • Availability
  • Add to Cart